Hexyl chloroformate 98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Hexyl carbonochloridate
427040050 427040250
ACRO427040050EA 79.6 EUR
ACRO427040050 ACRO427040250
Hexyl chloroformate 98%
Hexyl chloroformate
Formula: C₇H₁₃ClO₂
MW: 164.6299 g/mol
Boiling Pt: 60…61 °C (7 mmHg)
Flash Pt: 62 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000651
CAS Number: 6092-54-2
UN: 3277
ADR: 6.1 II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4230 to 1.4250 (20°C, 589 nm)
GC ≥97.5 %