Peqlab, Electroporation Cuvettes
Supplier: Avantor
Peqlab
732-2916EA 98.3 EUR
732-2916 732-2924 732-2918 732-2922 732-2920
Peqlab, Electroporation Cuvettes
Cuvettes Electroporation Cuvettes
High precision manufacturing and strict quality control guarantee consistent constant electroporation.
  • Coloured caps for selecting the correct column widths
  • 4 and 2 mm cuvettes for long electrodes
  • Sterilised with gamma rays
Balení: Individually packed to ensure sterility.