Safety helmets, uvex pheos B-WR
Supplier: UVEX
9772.930 9772.030 9772.530 9772.430 9772.230 9772.130 9772.330
111-1909EA 19.8 EUR
111-1909 111-1906 111-1879 111-1922 111-1921 111-1920 114-8685
Safety helmets, uvex pheos B-WR
Head Protection Safety Helmets
These ventilated helmets provide protection even in extremely low temperatures (down to −30 °C). They also provide protection against splashes of molten metal.
  • Three variable air vents aid airflow and help regulate the temperature within the helmet, making it more comfortable to wear
  • Adapter systems for attaching uvex visors, uvex earmuffs and lamps
  • Suspension harness with rotating ratchet wheel allows the width size to be adjusted for an individual fit
Size: 520 to 610 mm
EN 397, low temperature (–30 °C), and molten metal
Certifikace: EN 397,More Product Information