ESD safety shoes, lace-up, Velcro® fastening, uvex 1, 8516
Supplier: UVEX
8516.3/35 8516.3/36 8516.3/37 8516.3/38 8516.3/39 8516.3/40 8516.3/41 8516.3/42 8516.3/43 8516.3/44 8516.3/45 8516.3/46 8516.3/47 8516.3/48 8516.2/35 8516.2/36 8516.2/37 8516.2/38 8516.2/39 8516.2/40 8516.2/41 8516.2/42 8516.2/43 8516.2/44 8516.2/45 8516.2/46 8516.2/47 8516.2/48 8516.2/49 8516.2/50 8516.2/51 8516.2/52 8516.3/49 8516.3/50 8516.3/51 8516.3/52 8516.1/35 8516.1/36 8516.1/37 8516.1/38 8516.1/39 8516.1/40 8516.1/41 8516.1/42 8516.1/43 8516.1/51 8516.1/52 8516.1/44 8516.1/47 8516.1/48 8516.1/49 8516.1/50 8516.1/45 8516.1/46
114-7848EA 135 EUR
114-7848 114-7849 114-7850 114-7851 114-7852 114-7853 114-7854 114-7855 114-7856 114-7857 114-7858 114-7859 114-7860 114-7861 114-7273 114-7274 114-7275 114-7276 114-7277 114-7112 114-7113 114-7114 114-7115 114-7116 114-7117 114-7118 114-7119 114-7120 114-7121 114-7122 114-7123 114-7124 114-7862 114-7863 114-7864 114-7865 UVEX8516.1/35 UVEX8516.1/36 UVEX8516.1/37 UVEX8516.1/38 UVEX8516.1/39 UVEX8516.1/40 UVEX8516.1/41 UVEX8516.1/42 UVEX8516.1/43 UVEX8516.1/51 UVEX8516.1/52 UVEX8516.1/44 UVEX8516.1/47 UVEX8516.1/48 UVEX8516.1/49 UVEX8516.1/50 UVEX8516.1/45 UVEX8516.1/46
ESD safety shoes, lace-up, Velcro® fastening, uvex 1, 8516
Shoes and Boots
Metal-free, extremely lightweight, flexible and multifunctional safety shoe made from breathable water resistant high-tech micro velour upper, soft padded tongue and collar, plus a dual-density PUR sole (oil and petrol resistant, anti-static) free of lacquer-wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-slip sole with shock-absorption at heel and front of the foot
  • Virtually seamless design eliminates potential pressure points and blisters
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Flexible, metal-free, puncture resistant midsole
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black
Certifikace: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S3, SRC