Thermal socks, SK11
Supplier: Portwest
SK11BLU44-48 SK11BLU39-43 SK11GRR39-43 SK11GRR44-48
114-7156EA 7.3 EUR
114-7156 114-7155 114-7157 114-7158
Thermal socks, SK11
Workwear
These socks are made from 55% cotton, 15% nylon, 15% wool and 15% acrylic.
  • Designed for all seasons
  • Cushioning throughout provides extra warmth
  • Extra impact resistance and extra comfort
Height: 25 cm
Weight: 82 g