VWR®, Gardner colour standards
Supplier: VWR Chemicals
84815.260EA
208
EUR
84815.260
84816.260
84817.260
84818.260
84819.260
84820.260
84821.260
84822.260
VWR®, Gardner colour standards
Standards
Colour Reference and Spectroscopy Standards
Ready to use colour solutions that are prepared gravimetrically on a weight/weight basis that are designed for grading the colour of similarly coloured liquids, such as resins, varnishes, lacquers, drying oils, fatty acids, lecithin's, sunflower and linseed oil.
- Use either as calibration and/or quality control standards
- Designed specifically for use in ASTM analytical methods, and produced in accordance with ASTM D1500, D6045, D1209
- Traceable product certified by an independent laboratory
- Supplied in high quality, tamper-evident bottles