ESD safety ankle boots, lace-up, Safety-Grip, Renzo Biomex GTX
Supplier: ELTEN
765421/36 765421/37 765421/38 765421/39 765421/40 765421/41 765421/42 765421/43 765421/44 765421/45 765421/46 765421/47 765421/48 765421/49 765421/50
114-8191EA 125.9 EUR
114-8191 114-8192 114-8193 114-8194 114-8195 114-8196 114-8197 114-8198 114-8199 114-8200 114-8201 114-8202 114-8203 114-8204 114-8205
ESD safety ankle boots, lace-up, Safety-Grip, Renzo Biomex GTX
Shoes and Boots
Durable ESD safety shoes made from robust, breathable cowhide leather and hydrophobised nubuck leather, a full padded bellows tongue and a dual-density PU/PU sole safety grip (3,5 mm) with exceptional non-slip and good self-cleaning properties.
  • Gore-Tex® climate membrane
  • Replaceable insole; orthopaedic insoles possible
  • Full-length fleece inlay sole ESD PRO black
  • Anatomically shaped insole and ankle support for improved stability
  • Biomex-Protection®: Cuff stabilises ankles
  • Heat resistant seams and laces
  • PU scuff cap protects the leather upper
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in cold areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certifikace: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC, CI