ESD safety ankle boots, lace-up, Safety-Grip, Renzo XXW Mid
Supplier: ELTEN
765881/36 765881/37 765881/38 765881/39 765881/40 765881/41 765881/42 765881/43 765881/44 765881/45 765881/46 765881/47 765881/48 765881/49 765881/50
114-8176EA 68.9 EUR
114-8176 114-8177 114-8178 114-8179 114-8180 114-8181 114-8182 114-8183 114-8184 114-8185 114-8186 114-8187 114-8188 114-8189 114-8190
ESD safety ankle boots, lace-up, Safety-Grip, Renzo XXW Mid
Shoes and Boots
Extra wide durable ESD safety shoes made from robust, breathable cowhide leather, a full padded bellows tongue plus a dual-density PU/PU sole (Safety-Grip) (3,5 mm) with exceptional non slip and good self-cleaning properties.
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length fleece inlay sole (ESD Pro), black
  • Sole matches the movement of the foot, for more stability and less fatigue
  • Heat resistant seams and laces
  • PU scuff cap protects the leather upper
  • Puncture resistant steel midsole
  • Protective steel toecap (200 J)
  • Very wide version (XXW)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black
Certifikace: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC, Form B.