Diethyl chloromalonate 90%
Supplier: Thermo Fisher Scientific
Danger
114230050 114230250
ACRO114230050EA 77.2 EUR
ACRO114230050 ACRO114230250
Diethyl chloromalonate 90%
Diethyl chloromalonate
Formula: C₇H₁₁ClO₄
Melting Pt: 221…222 °C
Density: 1.204 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00009140
CAS Number: 14064-10-9
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless to pale yellow Liquid
Infrared spectrum Conforms
GC ≥88.0 %
HPLC ≥94 %