3-Methyl-2-pentanone 99%
Supplier: Thermo Scientific
Danger
127380050 127380250
ACRO127380050EA 55 EUR
ACRO127380050 ACRO127380250
3-Methyl-2-pentanone 99%
3-Methyl-2-pentanone
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 118 °C (758 mmHg)
Density: 0.81 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00009336
CAS Number: 565-61-7
UN: 1224
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥98.5 %
HPLC ≥96.0 %