Potassium fluoride 99.99% (trace metals basis)
Supplier: Thermo Fisher Scientific
Danger
146000050 146000250
ACRO146000050EA 82.8 EUR
ACRO146000050 ACRO146000250
Potassium fluoride 99.99% (trace metals basis)
Potassium fluoride
Formula: KF
Boiling Pt: 1505 °C (1013 hPa)
Melting Pt: 860 °C
Density: 2,48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011398
CAS Number: 7789-23-3
UN: 1812
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White Crystalline powder and/or chunks
Total trace metal impurities ≤100 ppm
GC ≥96.0 %