3-Methoxy-2(1H)-pyridone 97%
Supplier: Thermo Scientific
Warning
Synonyms: 2-Hydroxy-3-methoxypyridine
163540010 163540050
ACRO163540010EA 77.2 EUR
ACRO163540010 ACRO163540050
3-Methoxy-2(1H)-pyridone 97%
3-Methoxy-2(1H)-pyridone
Formula: C₆H₇NO₂
MW: 125.1253 g/mol
Melting Pt: 113…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00006015
CAS Number: 20928-63-6
Order Now
Specification Test Results
Appearance White to brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %