Cesium iodide 99.999% (trace metals basis)
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Cesiumiodide
192820010 192820100 192820500
ACRO192820010EA 30.4 EUR
ACRO192820010 ACRO192820100 ACRO192820500
Cesium iodide 99.999% (trace metals basis)
Cesium iodide
Formula: CsI
MW: 259.81 g/mol
Boiling Pt: 1280 °C (760 mmHg)
Melting Pt: 626 °C
Storage Temperature: Ambient
MDL Number: MFCD00010962
CAS Number: 7789-17-5
UN: 3077
ADR: 9 III
Merck Index: 12,02056
Order Now
Specification Test Results
Appearance White Crystalline powder
Total trace metal impurities ≤10 ppm