Magnesium chloride, anhydrous 99.9% (trace metals basis), pure
Supplier: Thermo Fisher Scientific
197060100 197060500
ACRO197060100EA 129 EUR
ACRO197060100 ACRO197060500
Magnesium chloride, anhydrous 99.9% (trace metals basis), pure
Magnesium chloride
Formula: MgCl₂
MW: 95.21 g/mol
Boiling Pt: 1412 °C (760 mmHg)
Melting Pt: 708 °C
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
Merck Index: 13,05684
Order Now
Specification Test Results
Appearance White Powder to beads
Total trace metal impurities ≤0.1 %