Calcium chloride ACS desiccant
Supplier: Thermo Fisher Scientific
Warning
206791000 206795000
ACRO206791000EA 52.1 EUR
ACRO206791000 ACRO206795000
Calcium chloride ACS desiccant
Calcium chloride
Formula: CaCl₂
Boiling Pt: >1600 °C (760 mmHg)
Melting Pt: 782 °C
Density: 2,174 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
Order Now
Specification Test Results
Appearance White Powder
Titration Complexometric ≥96.0 %
Titrable base ≤0.006 meq/g