Alloxan monohydrate 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Pyrimidine-2,4,5,6(1H,3H)-tetraone hydrate
215370250 215370100 215371000
ACRO215370250EA 75.7 EUR
ACRO215370250 ACRO215370100 ACRO215371000
Alloxan monohydrate 98%
Alloxan monohydrate
Formula: C₄H₄N₂O₅
MW: 160.09 g/mol
Melting Pt: 240 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00006031
CAS Number: 2244-11-3
Order Now
Specification Test Results
Appearance Off-white to yellow to light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %