Lithiumdiisopropylamide 2 M in THF/n-heptane/ethylbenzene, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
268831000 268838000
268831000.EA 69.1 EUR
268831000. 268838000.
Lithiumdiisopropylamide 2 M in THF/n-heptane/ethylbenzene, AcroSeal®
Lithiumdiisopropylamide
Formula: C₆H₁₄LiN
Boiling Pt: 60…136 °C (760 mmHg)
Density: 0.81 g/cm³
Flash Pt: 2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00064449
CAS Number: 4111-54-0
UN: 2924
ADR: 3,II
Order Now
Specification Test Results
Appearance Dark yellow to orange to dark red-brown Solution
Additional info Remark: keep soln. at < 10°C at which it is stable
Titration with HClO4 24.6 to 30.0 %