2,3-Naphthalenedialdehyde (NDA) 95%

Supplier: Thermo Scientific
Warning

Synonyms: Naphthalene-2,3-dicarbaldehyde, Naphthalene-2,3-dicarboxaldehyde, NDA

294721000 294725000
ACRO294721000EA 131 EUR
ACRO294721000 ACRO294725000
2,3-Naphthalenedialdehyde (NDA) 95%
2,3-Naphthalenedialdehyde (NDA)
Formula: C₁₂H₈O₂
MW: 184.19 g/mol
Melting Pt: 134…135 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00016618
CAS Number: 7149-49-7

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR