Trimethyl borate 99%, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
346121000 346128000
346121000.EA 39.7 EUR
346121000. 346128000.
Trimethyl borate 99%, AcroSeal®
Trimethyl borate
Formula: C₃H₉BO₃
MW: 103.91 g/mol
Boiling Pt: 68…69 °C (760 mmHg)
Melting Pt: –34 °C
Density: 0.915 g/cm³
Flash Pt: –8 °C
Storage Temperature: Ambient
MDL Number: MFCD00008346
CAS Number: 121-43-7
UN: 2416
ADR: 3 II
Merck Index: 12,09842
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %