(R)-(+)-2-Methyl-CBS-oxazaborolidine 1 M in toluene
Supplier: Thermo Fisher Scientific
Danger
Synonyms: (R)-Tetrahydro-1-methyl-3,3-diphenyl-1H,3H-pyrrolo[1,2-c][1,3,2]oxazaborole
369780250 369780050
ACRO369780250EA 269 EUR
ACRO369780250 ACRO369780050
(R)-(+)-2-Methyl-CBS-oxazaborolidine 1 M in toluene
(R)-(+)-2-Methyl-CBS-oxazaborolidine
Formula: C₁₈H₂₀BNO
MW: 277.17 g/mol
Boiling Pt: 111 °C (760 mmHg)
Density: 0.95 g/cm³
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00078440
CAS Number: 112022-83-0
UN: 1294
ADR: 3 II
Order Now
Specification Test Results
Appearance Colorless to amber Solution
Strength 0.90 to 1.10 M