5-Bromo-1-methylimidazole 97%
Supplier: Thermo Scientific
Danger
Synonyms: 5-Bromo-1-methyl-1H-imidazole
381040050 381040250
ACRO381040050EA 141 EUR
ACRO381040050 ACRO381040250
5-Bromo-1-methylimidazole 97%
5-Bromo-1-methylimidazole
Formula: C₄H₅BrN₂
MW: 161 g/mol
Boiling Pt: 120 °C (15 mmHg)
Melting Pt: 40…41 °C
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD01632218
CAS Number: 1003-21-0
UN: 2923
ADR: 8,II
Order Now
Specification Test Results
Appearance White to tan Crystalline solid
Infrared spectrum Conforms
Melting point 38 to 43 °C
GC ≥96.0 %