1(2H)-Phthalazinone 98+%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Phthalazone, Phthalazin-1(2H)-one

384570050 384570250 384571000 384575000
ACRO384570050EA 28.8 EUR
ACRO384570050 ACRO384570250 ACRO384571000 ACRO384575000
1(2H)-Phthalazinone 98+%
1(2H)-Phthalazinone
Formula: C₈H₆N₂O
Melting Pt: 184…187 °C
Storage Temperature: Ambient
MDL Number: MFCD00006892
CAS Number: 119-39-1

Order Now

Specification Test Results

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 184 to 187 °C
HPLC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR