Samarium(III) trifluoromethanesulphonate 98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Samarium(III) trifluoromethanesulphonate
383170010 383170100
ACRO383170010EA 26.2 EUR
ACRO383170010 ACRO383170100
Samarium(III) trifluoromethanesulphonate 98%
Samarium (III) trifluoromethanesulphonate
Formula: C₃F₉O₉S₃Sm
MW: 597.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00192431
CAS Number: 52093-28-4
UN: 3261
ADR: 8 III
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Elemental Analysis 24.65 to 25.66 % (Sm)