Pumice stone, granular (∼ 1-5 mm)
Supplier: Thermo Fisher Scientific
390040010 390042500
ACRO390040010EA 102 EUR
ACRO390040010 ACRO390042500
Pumice stone, granular (∼ 1-5 mm)
Pumice stone
Storage Temperature: Ambient
MDL Number: MFCD00148147
CAS Number: 1332-09-8
Order Now
Specification Test Results
Appearance Grey-brown Granules
Particle size ≥90 % (0.8 - 3 mm)