Indole-5-carbaldehyd 98+%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Indole-5-carboxaldehyde, 1H-Indole-5-carboxaldehyde
391630010 391630050 391630250
ACRO391630010EA 37.4 EUR
ACRO391630010 ACRO391630050 ACRO391630250
Indole-5-carbaldehyd 98+%
Indole-5-carbaldehyd
Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 100…103 °C
Storage Temperature: Ambient
MDL Number: MFCD02093664
CAS Number: 1196-69-6
Order Now
Specification Test Results
Appearance Orange to tan Powder
Infrared spectrum Conforms
Melting point 98 to 103 °C
HPLC ≥98.0 %