Trisodium citrate, anhydrous 98%, pure
Supplier: Thermo Fisher Scientific
Synonyms: Sodium Citrate, Sodium citrate tribasic, Citric acid trisodium salt, Sodium 2-hydroxypropane-1,2,3-tricarboxylate
391975000 391970025 391971000
ACRO391975000EA 67.7 EUR
ACRO391975000 ACRO391970025 ACRO391971000
Trisodium citrate, anhydrous 98%, pure
tri-Sodium citrate
Formula: C₆H₅Na₃O₇
MW: 258.07 g/mol
Melting Pt: 300 °C
Flash Pt: 290 °C
Storage Temperature: Ambient
MDL Number: MFCD00012462
CAS Number: 68-04-2
Merck Index: 12,08746
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %