4-(Hydroxymethyl)benzoic acid 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 4-Carboxybenzylalcohol, HMBA Linker, HMBA
394340010 394340100
ACRO394340010EA 54.5 EUR
ACRO394340010 ACRO394340100
4-(Hydroxymethyl)benzoic acid 98%
4-(Hydroxymethyl)benzoic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 182…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00017598
CAS Number: 3006-96-0
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 181 to 186 °C
HPLC ≥97.5 %