2-Methylnicotinic acid 98+%
Supplier: Thermo Scientific
Warning
Synonyms: 2-Methylpyridine-3-carboxylic acid, 2-Methyl-3-pyridinecarboxylic acid
397320010 397320050 397320250
ACRO397320010EA 77.5 EUR
ACRO397320010 ACRO397320050 ACRO397320250
2-Methylnicotinic acid 98+%
2-Methylnicotinic acid
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: 225…228 °C
Storage Temperature: Ambient
MDL Number: MFCD00013449
CAS Number: 3222-56-8
Order Now
Specification Test Results
Appearance Off-white to light brown Powder
Infrared spectrum Conforms
HPLC ≥98.0 %
Titration with NaOH 97.5 to 102.5 %