Cyclopentane 95+%, pure
Supplier: Thermo Scientific
Danger
Synonyms: Pentamethylene
397430010 397430050
ACRO397430010EA 105 EUR
ACRO397430010 ACRO397430050
Cyclopentane 95+%, pure
Cyclopentane
Formula: C₅H₁₀
MW: 70.13 g/mol
Boiling Pt: 50 °C (760 mmHg)
Melting Pt: –94 °C
Density: 0.75 g/cm³
Flash Pt: –7 °C
Storage Temperature: Ambient
MDL Number: MFCD00001356
CAS Number: 287-92-3
UN: 1146
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4dd230057521cc77c90840c7adfece9c74c0a3b1&vid=352021851b7701a448c5b11d43e94d925d03f802&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥95.0 %