Methyl indole-6-carboxylate 97%
Supplier: Thermo Scientific
Warning
Synonyms: Methyl 1H-indole-6-carboxylate
397930010 397930050
ACRO397930010EA 29.4 EUR
ACRO397930010 ACRO397930050
Methyl indole-6-carboxylate 97%
Methyl indole-6-carboxylate
Formula: C₁₀H₉NO₂
MW: 175.19 g/mol
Melting Pt: 76…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00211063
CAS Number: 50820-65-0
Order Now
Specification Test Results
Appearance White to light brown Powder
Infrared spectrum Conforms
Melting point 79 to 84 °C
GC ≥96.0 %