6-Fluoroveratraldehyde 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2-Fluoro-4,5-dimethoxybenzaldehyde, 4,5-Dimethoxy-2-fluorobenzaldehyde, 3,4-Dimethoxy-6-fluorobenzaldehyde
398090010 398090050
ACRO398090010EA 40.2 EUR
ACRO398090010 ACRO398090050
6-Fluoroveratraldehyde 97%
6-Fluoroveratraldehyde
Formula: C₉H₉FO₃
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00061108
CAS Number: 71924-62-4
Order Now
Specification Test Results
Appearance White to light yellow Powder or solid
Infrared spectrum Conforms
Melting point 98 to 100 °C
GC ≥96.0 %