Ethyl (4-bromobenzoyl)acetate 95%
Supplier: Thermo Scientific
Warning
Synonyms: ethyl 3-(4-bromophenyl)-3-oxopropanoate, 3-(4-bromophenyl)-3-oxo-propionic acid ethyl ester, Benzenepropanoic acid, 4-bromo-beta-oxo-, ethyl ester
399160010 399160050
ACRO399160010EA 95.4 EUR
ACRO399160010 ACRO399160050
Ethyl (4-bromobenzoyl)acetate 95%
Ethyl (4-bromobenzoyl)acetate
Formula: C₁₁H₁₁BrO₃
MW: 271.11 g/mol
Boiling Pt: 268…269 °C (760 mmHg)
Density: 1.432 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00231243
CAS Number: 26510-95-2
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
HPLC ≥94 %