Pyridazine-4-carboxylic acid 95%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 4-Pyridazinecarboxylic acid
399630010 399630050
ACRO399630010EA 72.3 EUR
ACRO399630010 ACRO399630050
Pyridazine-4-carboxylic acid 95%
Pyridazine-4-carboxylic acid
Formula: C₅H₄N₂O₂
MW: 124.10 g/mol
Melting Pt: 244.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00011576
CAS Number: 50681-25-9
Order Now
Specification Test Results
Appearance Off-white to brown Powder
Infrared spectrum Conforms
HPLC ≥94 %