o-Cresol 99%, AcroSeal®
Supplier: Thermo Scientific
Danger
Synonyms: 1-Hydroxy-2-methylbenzene, 2-Methylphenol, 2-Hydroxytoluene
405731000 405738000
405731000.EA 20.5 EUR
405731000. 405738000.
o-Cresol 99%, AcroSeal®
o-Cresol
Formula: C₇H₈O
MW: 108.14 g/mol
Boiling Pt: 191 °C (760 mmHg)
Melting Pt: 30…32 °C
Density: 1.04 g/cm³
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00002226
CAS Number: 95-48-7
UN: 3455
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Colorless to amber Low melting solid or liquid (May darken on storage)
Infrared spectrum Conforms
Water ≤0.2 %
GC ≥98.5 %