Methyl-1,2,4-triazole-3-carboxylate 98%
Supplier: Thermo Scientific
Warning
Synonyms: Methyl 1H-1,2,4-triazole-3-carboxylate, Methyl 1H-1,2,4-triazole-5-carboxylate
415000050 415000250
ACRO415000050EA 35 EUR
ACRO415000050 ACRO415000250
Methyl-1,2,4-triazole-3-carboxylate 98%
Methyl 1,2,4-triazole-3-carboxylate
Formula: C₄H₅N₃O₂
MW: 127.10 g/mol
Melting Pt: 190…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00135989
CAS Number: 4928-88-5
Order Now
Specification Test Results
Appearance White Powder and/or flakes
Infrared spectrum Conforms
Melting point 185 to 193 °C
HPLC ≥97.5 %