Ethyl glyoxylate ∼50% in toluene, Technical Grade
Supplier: Thermo Scientific
Danger
425760010 425765000
ACRO425760010EA 230 EUR
ACRO425760010 ACRO425765000
Ethyl glyoxylate ∼50% in toluene, Technical Grade
Ethyl glyoxylate
Formula: C₄H₆O₃
MW: 102.09 g/mol
Boiling Pt: 110 °C (760 mmHg)
Density: 1.032 g/cm³
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00044009
CAS Number: 924-44-7
UN: 1294
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Solution
Titration excess HCl ≥48 % (Aldehyde function)