(3-Acryloxypropyl)trimethoxysilane 90% stabilised
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 3-(Trimethoxysilyl)propyl acrylate
426600050 426600250
ACRO426600050EA 106 EUR
ACRO426600050 ACRO426600250
(3-Acryloxypropyl)trimethoxysilane 90% stabilised
(3-Acryloxypropyl)trimethoxysilane
Formula: C₉H₁₈O₅Si
MW: 234.32 g/mol
Boiling Pt: 95 °C (3 mmHg)
Density: 1.059 g/cm³
Flash Pt: 123 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00054803
CAS Number: 4369-14-6
UN: 3265
ADR: 8 III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4280 to 1.4300 (20°C, 589 nm)
GC ≥90 %
Stabilizer Stabilized with BHT