5,6-Dihydro-2H-pyran-2-one 90%
Supplier: Thermo Fisher Scientific
Warning
426640010 426640050 426640250
ACRO426640010EA 64.4 EUR
ACRO426640010 ACRO426640050 ACRO426640250
5,6-Dihydro-2H-pyran-2-one 90%
5,6-Dihydro-2H-pyran-2-one
Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 103 °C (10 mmHg)
Density: 1.139 g/cm³
Flash Pt: 46 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010439
CAS Number: 3393-45-1
UN: 1224
ADR: 3 III
Order Now
Specification Test Results
Appearance Colorless to yellow to orange Liquid
Infrared spectrum Conforms
GC ≥96.0 %