p-Tolylmagnesium bromide ∼0.5 M in diethyl ether, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
Synonyms: p-Tolylmagnesium bromide
426761000 426768000
426761000.EA 57.1 EUR
426761000. 426768000.
p-Tolylmagnesium bromide ∼0.5 M in diethyl ether, AcroSeal®
p-Tolylmagnesium bromide
Formula: C₇H₇BrMg
Density: 0.866 g/cm³
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00000040
CAS Number: 4294-57-9
UN: 2924
ADR: 3,I
Order Now
Specification Test Results
Appearance Clear to hazy yellow to amber Solution (May develop some turbidity or precipitate)
Titration Grignard 0.40 to 0.60 M
Rhodium (Rh) ≥34.7 %