Phosphorus(V) bromide 95%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Phosphorus pentabromide
428270250 428271000
ACRO428270250EA 83.3 EUR
ACRO428270250 ACRO428271000
Phosphorus(V) bromide 95%
Phosphorus (V) bromide
Formula: PBr₅
MW: 430.49 g/mol
Melting Pt: 105…107 °C (dec.)
Density: 3.61
Storage Temperature: Ambient
MDL Number: MFCD00011437
CAS Number: 7789-69-7
UN: 2691
ADR: 8 II
Merck Index: 12,07507
Order Now
Specification Test Results
Appearance Yellow to orange to red Crystalline powder or crystals and/or chunks
Titration Argentometric ≥94 %
Insoluble matter Pass