Trimethyl orthoformate, anhydrous 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
429281000 429288000
429281000.EA 39.6 EUR
429281000. 429288000.
Trimethyl orthoformate, anhydrous 99+%, AcroSeal®
Trimethyl orthoformate
Formula: C₄H₁₀O₃
MW: 106.12 g/mol
Boiling Pt: 101…102 °C (760 mmHg)
Melting Pt: –53 °C
Density: 0.97 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00008483
CAS Number: 149-73-5
UN: 3271
ADR: 3 II
Merck Index: 12,07012
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %
Residue after evaporation ≤0.005 %