Amberlite® IRN-78, Ion exchange resin cationic type (OH-) resin_ion-exchange
Supplier: Thermo Fisher Scientific
Danger
429290025 429291000 429295000
ACRO429290025EA 286 EUR
ACRO429290025 ACRO429291000 ACRO429295000
Amberlite® IRN-78, Ion exchange resin cationic type (OH-) resin_ion-exchange
AMBERLITE® IRN-78, Ion exchange resin cationic type (OH-)
Storage Temperature: Ambient
MDL Number: MFCD00145822
CAS Number: 11128-95-3
Order Now
Specification Test Results
Appearance Yellow to amber Spherical beads
Harmonic mean size 0.580 to 0.680 mm
Uniformity coefficient ≤1.20
Moisture holding capacity 54.0 to 60.0 % (OH)
Total volume capacity ≥1.10 eq/l (OH) as is