4-Bromophenyl chloroformate 95+%
Supplier: Thermo Fisher Scientific
Danger
430150010 430150050
ACRO430150010EA 34.9 EUR
ACRO430150010 ACRO430150050
4-Bromophenyl chloroformate 95+%
4-Bromophenyl chloroformate
Formula: C₇H₄BrClO₂
Boiling Pt: 118 °C (16hPa)
Density: 1.648 g/cm³
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013256
CAS Number: 7693-44-9
UN: 3277
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Low melting solid or liquid
Infrared spectrum Conforms
GC ≥97.5 %