2-Nitrophenylboronic acid 97%
Supplier: Thermo Scientific
Warning
Synonyms: Dihydroxy(2-nitrophenyl)borane, (2-Nitrophenyl)boronic acid, 2-Nitrobenzeneboronic acid, (2-Nitrophenyl)metaboric acid
430590010 430590050
ACRO430590010EA 69 EUR
ACRO430590010 ACRO430590050
2-Nitrophenylboronic acid 97%
2-Nitrophenylboronic acid
Formula: C₆H₆BNO₄
MW: 166.93 g/mol
Melting Pt: 147…152 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00161358
CAS Number: 5570-19-4
Order Now
Specification Test Results
Appearance Brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥98.5 %