4-(Methoxycarbonyl)phenylboronic acid pinacol ester 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 4-(Methoxycarbonyl)benzeneboronic acid pinacol ester, Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
430610010 430610050
ACRO430610010EA 48 EUR
ACRO430610010 ACRO430610050
4-(Methoxycarbonyl)phenylboronic acid pinacol ester 97%
4-(Methoxycarbonyl)phenylboronic acid pinacol ester
Formula: C₁₄H₁₉BO₄
MW: 262.11 g/mol
Melting Pt: 82…83.2 °C
Storage Temperature: Ambient
MDL Number: MFCD02179438
CAS Number: 171364-80-0
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 81 to 84 °C
GC ≥96.0 %