2-Chloro-3-pyridineboronic acid pinacol ester 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 2-Chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-Chloropyridine-3-boronic acid pinacol ester
430660010 430660050
ACRO430660010EA 24.9 EUR
ACRO430660010 ACRO430660050
2-Chloro-3-pyridineboronic acid pinacol ester 97%
2-Chloro-3-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BClNO₂
MW: 239.51 g/mol
Melting Pt: 34…39 °C
Storage Temperature: Ambient
MDL Number: MFCD04039873
CAS Number: 452972-11-1
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %
HPLC ≥97.0 %