kappa-Carrageenan
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Irish moss
431590250 431591000
ACRO431590250EA 40.2 EUR
ACRO431590250 ACRO431591000
kappa-Carrageenan
kappa-Carrageenan
Formula: C₁₂H₁₇O₁₃S
MW: 401.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151514
CAS Number: 11114-20-8
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Appearance White to off-white to yellow to brown Powder
Infrared spectrum Conforms
Loss on drying ≤12 % (1 g, 105°C)
Viscosity ≥10 mPa.s (0.3 % in water at 25°C)
Ash ≤40 %
Barium (Ba) ≤0.005 %
Calcium (Ca) ≤0.001 %
Lithium (Li) ≤0.001 %
Magnesium (Mg) ≤0.0002 %
Potassium (K) ≤0.03 %
Rubidium (Rb) ≤0.005 %
Sodium (Na) ≤0.02 %
Strontium (Sr) ≤0.0005 %