3-Quinolineboronic acid pinacol ester 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Quinoline-3-boronic acid pinacolate, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline, Quinoline-3-boronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-(3-quinolyl)-1,3,2-dioxaborolane
432110010 432110050
ACRO432110010EA 65.1 EUR
ACRO432110010 ACRO432110050
3-Quinolineboronic acid pinacol ester 97%
3-Quinolineboronic acid pinacol ester
Formula: C₁₅H₁₈BNO₂
MW: 255.1199 g/mol
Melting Pt: 55…57 °C
Storage Temperature: Ambient
MDL Number: MFCD05155218
CAS Number: 171364-85-5
Order Now
Specification Test Results
Appearance White to light brown Powder or solid
Infrared spectrum Conforms
Melting point 55 to 60 °C
GC ≥96.0 %