4-Nitrophenylboronic acid 97%
Supplier: Thermo Scientific
Warning
Synonyms: 4-Nitrobenzeneboronic acid, (4-Nitrophenyl)boronic acid, Dihydroxy(4-nitrophenyl)borane, (4-Nitrophenyl)metaboric acid
433890010 433890050
ACRO433890010EA 61.2 EUR
ACRO433890010 ACRO433890050
4-Nitrophenylboronic acid 97%
4-Nitrophenylboronic acid
Formula: C₆H₆BNO₄
MW: 166.93 g/mol
Melting Pt: 270…271 °C
Storage Temperature: Ambient
MDL Number: MFCD00161360
CAS Number: 24067-17-2
Order Now
Specification Test Results
Appearance White to brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %