6-Hydroxypicolinic acid 95%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 6-Hydroxy-2-pyridinecarboxylic acid, 6-Hydroxypyridine-2-carboxylic acid
434820050 434820250
ACRO434820050EA 51.2 EUR
ACRO434820050 ACRO434820250
6-Hydroxypicolinic acid 95%
6-Hydroxypicolinic acid
Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 273…276 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00192220
CAS Number: 19621-92-2
Order Now
Specification Test Results
Appearance White to yellow to brown Powder
Infrared spectrum Conforms
HPLC ≥98.5 %